Assay Details
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062822.1 | XP_017918311.1 | 2 - 3 | 456 | 56 | |
XM_018062823.1 | XP_017918312.1 | 2 - 3 | 2803 | 56 | ||
XM_018062825.1 | XP_017918314.1 | 2 - 3 | 901 | 56 | ||
XM_018062826.1 | XP_017918315.1 | 2 - 3 | 603 | 56 | ||
GenBank mRNA | KA338826.1 | - | 3 - 4 | 1255 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
