Assay Details
| Gene Symbol: ADCYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039817.1 | XP_017895306.1 | 3 - 4 | 872 | 73 | |
| GenBank mRNA | JO630837.2 | - | 2 - 3 | 589 | 73 | |
More Information
Set Membership: |
|

ADCYAP1
Genomic Map