Assay Details
Gene Symbol: RCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040887.1 | XP_017896376.1 | 3 - 4 | 726 | 60 | |
GenBank mRNA | JO430308.2 | - | 8 - 9 | 1417 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
