Assay Details
Gene Symbol: TFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675635.3 | XP_005675692.1 | 2 - 3 | 293 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
