Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677834.2 | XP_005677891.1 | 5 - 6 | 1290 | 87 | |
XM_005677835.3 | XP_005677892.1 | 5 - 6 | 709 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
