Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686851.3 | XP_005686908.1 | 2 - 3 | 229 | 97 | |
XM_013967823.2 | XP_013823277.1 | 2 - 3 | 441 | 97 | ||
XM_018055481.1 | XP_017910970.1 | 2 - 3 | 462 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
