Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039236.1 | XP_017894725.1 | 6 - 7 | 1148 | 61 | |
| XM_018039237.1 | XP_017894726.1 | 6 - 7 | 1047 | 61 | ||
| XM_018039238.1 | XP_017894727.1 | 6 - 7 | 1171 | 61 | ||
| XM_018039239.1 | XP_017894728.1 | 6 - 7 | 867 | 61 | ||
More Information
Set Membership: |
|

PACSIN1
Genomic Map