Assay Details
Gene Symbol: ABHD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697786.3 | XP_005697843.1 | 1 - 2 | 306 | 68 | |
XM_005697788.3 | XP_005697845.1 | 1 - 2 | 188 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
