Assay Details
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963597.2 | XP_013819051.1 | 8 - 9 | 1300 | 56 | |
GenBank mRNA | JO499336.2 | - | 4 - 5 | 499 | 56 | |
JO500738.2 | - | 4 - 5 | 499 | 56 | ||
JO614942.2 | - | 8 - 9 | 1486 | 56 | ||
JT705915.1 | - | 2 - 3 | 235 | 56 | ||
JT717633.1 | - | 2 - 3 | 224 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
