Assay Details
Gene Symbol: NKX3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966088.2 | XP_013821542.2 | 1 - 2 | 391 | 69 | |
GenBank mRNA | JT660726.1 | - | 7 - 8 | 1171 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
