Assay Details
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062744.1 | XP_017918233.1 | 4 - 5 | 461 | 62 | |
XM_018062745.1 | XP_017918234.1 | 3 - 4 | 422 | 62 | ||
GenBank mRNA | JO411284.2 | - | 4 - 5 | 432 | 62 | |
JO434791.2 | - | 5 - 6 | 730 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
