Assay Details
Gene Symbol: GCGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063580.1 | XP_017919069.1 | 10 - 11 | 3944 | 61 | |
XM_018063581.1 | XP_017919070.1 | 11 - 12 | 1415 | 61 | ||
XM_018063582.1 | XP_017919071.1 | 11 - 12 | 1308 | 61 | ||
XM_018063584.1 | XP_017919073.1 | 12 - 13 | 1345 | 61 | ||
XM_018063585.1 | XP_017919074.1 | 9 - 10 | 3776 | 61 | ||
XM_018063586.1 | XP_017919075.1 | 9 - 10 | 960 | 61 | ||
GenBank mRNA | JO421548.2 | - | 10 - 11 | 1033 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
