Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042002.1 | XP_017897491.1 | 2 - 3 | 883 | 64 | |
XM_018042003.1 | XP_017897492.1 | 2 - 3 | 883 | 64 | ||
GenBank mRNA | JO426820.1 | - | 2 - 3 | 878 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
