Assay Details
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676330.3 | XP_005676387.1 | 1 - 2 | 123 | 66 | |
GenBank mRNA | JT694230.1 | - | 11 - 12 | 2130 | 66 | |
KA330340.1 | - | 1 - 2 | 137 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
