Assay Details
Gene Symbol: LOC102188370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677751.2 | XP_005677808.1 | 10 - 11 | 1061 | 84 | |
XM_018046005.1 | XP_017901494.1 | 8 - 9 | 762 | 84 | ||
GenBank mRNA | JO594747.2 | - | 10 - 11 | 1020 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
