Assay Details
Gene Symbol: LOC102188370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677751.2 | XP_005677808.1 | 13 - 14 | 1313 | 78 | |
XM_005677753.3 | XP_005677810.1 | 12 - 13 | 1218 | 78 | ||
XM_018046005.1 | XP_017901494.1 | 11 - 12 | 1014 | 78 | ||
GenBank mRNA | JO594747.2 | - | 13 - 14 | 1272 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
