Assay Details
Gene Symbol: LOC102188370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677751.2 | XP_005677808.1 | 9 - 10 | 970 | 132 | |
XM_005677753.3 | XP_005677810.1 | 9 - 10 | 971 | 132 | ||
XM_018046005.1 | XP_017901494.1 | 7 - 8 | 671 | 132 | ||
GenBank mRNA | JO594747.2 | - | 9 - 10 | 929 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
