Assay Details
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678522.2 | XP_005678579.2 | 9 - 10 | 2248 | 58 | |
XM_005678523.3 | XP_005678580.2 | 9 - 10 | 2182 | 58 | ||
XM_005678524.3 | XP_005678581.2 | 9 - 10 | 2177 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
