Assay Details
| Gene Symbol: LOC102186672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049509.1 | XP_017904998.1 | 3 - 4 | 476 | 93 | |
| XM_018049510.1 | XP_017904999.1 | 3 - 4 | 475 | 93 | ||
| XM_018049511.1 | XP_017905000.1 | 3 - 4 | 437 | 93 | ||
More Information
Set Membership: |
|

LOC102186672
Genomic Map