Assay Details
| Gene Symbol: LOC102186672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049509.1 | XP_017904998.1 | 7 - 8 | 924 | 75 | |
| XM_018049510.1 | XP_017904999.1 | 6 - 7 | 884 | 75 | ||
| XM_018049511.1 | XP_017905000.1 | 7 - 8 | 885 | 75 | ||
More Information
Set Membership: |
|

LOC102186672
Genomic Map