Assay Details
Gene Symbol: NMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686290.2 | XP_005686347.2 | 5 - 6 | 350 | 64 | |
GenBank mRNA | GQ150540.1 | - | 5 - 6 | 196 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
