Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693858.3 | XP_005693915.2 | 3 - 4 | 501 | 54 | |
GenBank mRNA | JO426162.1 | - | 3 - 4 | 581 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
