Assay Details
| Gene Symbol: TRUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041655.1 | XP_017897144.1 | 5 - 6 | 647 | 79 | |
| XM_018041656.1 | XP_017897145.1 | 5 - 6 | 644 | 79 | ||
| GenBank mRNA | JT692043.1 | - | 5 - 6 | 574 | 79 | |
More Information
Set Membership: |
|

TRUB1
Genomic Map