Assay Details
| Gene Symbol: TMPRSS11D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964668.2 | XP_013820122.1 | 7 - 8 | 810 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11D| Gene Symbol: TMPRSS11D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964668.2 | XP_013820122.1 | 7 - 8 | 810 | 66 | |
Set Membership: |
|