Assay Details
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056585.1 | XP_017912074.1 | 6 - 7 | 976 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
