Assay Details
| Gene Symbol: ASPHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061066.1 | XP_017916555.1 | 1 - 2 | 1095 | 73 | |
| GenBank mRNA | JO624403.2 | - | 2 - 3 | 211 | 73 | |
More Information
Set Membership: |
|

ASPHD2
Genomic Map