Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063043.1 | XP_017918532.1 | 3 - 4 | 841 | 102 | |
| XM_018063044.1 | XP_017918533.1 | 3 - 4 | 736 | 102 | ||
| XM_018063045.1 | XP_017918534.1 | 3 - 4 | 281 | 102 | ||
| GenBank mRNA | KA310395.1 | - | 3 - 4 | 738 | 102 | |
More Information
Set Membership: |
|

CCDC106
Genomic Map