Assay Details
Gene Symbol: LOC108635132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045407.1 | XP_017900896.1 | 377 | 114 | ||
GenBank mRNA | JO413457.2 | - | 742 | 114 |
Gene Symbol: DIAPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO593540.2 | - | 2460 | 114 |

More Information
Set Membership: |
![]() ![]() |
