Assay Details
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044592.1 | XP_017900081.1 | 2 - 3 | 268 | 57 | |
XM_018044593.1 | XP_017900082.1 | 2 - 3 | 268 | 57 | ||
XM_018044594.1 | XP_017900083.1 | 2 - 3 | 268 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
