Assay Details
| Gene Symbol: PKLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976777.2 | XP_013832231.2 | 10 - 11 | 1626 | 63 | |
| XM_018046254.1 | XP_017901743.1 | 9 - 10 | 2107 | 63 | ||
| XM_018046255.1 | XP_017901744.1 | 10 - 11 | 1584 | 63 | ||
More Information
Set Membership: |
|

PKLR
Genomic Map