Assay Details
| Gene Symbol: LOC102169515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO380579.2 | - | 3 - 4 | 332 | 67 | |
| JO530475.2 | - | 1 - 2 | 185 | 67 | ||
| JO534269.2 | - | 3 - 4 | 358 | 67 | ||
| JO576862.1 | - | 3 - 4 | 410 | 67 | ||
| JO638824.2 | - | 3 - 4 | 341 | 67 | ||
| JT705727.1 | - | 1 - 2 | 165 | 67 | ||
| JT737790.1 | - | 1 - 2 | 188 | 67 | ||
| KA344759.1 | - | 3 - 4 | 331 | 67 | ||
| Gene Symbol: SMIM11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056532.1 | XP_017912021.1 | 3 - 4 | 374 | 67 | |
| GenBank mRNA | JT658372.1 | - | 2 - 3 | 241 | 67 | |
More Information
Set Membership: |
|

LOC102169515
Genomic Map