Assay Details
Gene Symbol: CAVIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676284.3 | XP_005676341.2 | 1 - 2 | 817 | 63 | |
GenBank mRNA | JI861874.1 | - | 1 - 2 | 484 | 63 | |
JO559564.2 | - | 1 - 2 | 1251 | 63 | ||
JT677763.1 | - | 4 - 5 | 2173 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
