Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060260.1 | XP_017915749.1 | 7 - 8 | 995 | 74 | |
XM_018060270.1 | XP_017915759.1 | 5 - 6 | 844 | 74 | ||
GenBank mRNA | JI861886.1 | - | 7 - 8 | 649 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
