Assay Details
Gene Symbol: SPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680514.3 | XP_005680571.1 | 5 - 6 | 416 | 68 | |
GenBank mRNA | JO581258.1 | - | 5 - 6 | 467 | 68 | |
JT687080.1 | - | 4 - 5 | 342 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
