Assay Details
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680937.3 | XP_005680994.1 | 2 - 3 | 935 | 68 | |
XM_018048550.1 | XP_017904039.1 | 2 - 3 | 892 | 68 | ||
GenBank mRNA | JO436962.2 | - | 2 - 3 | 918 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
