Assay Details
| Gene Symbol: PLVAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682121.3 | XP_005682178.2 | 3 - 4 | 1278 | 55 | |
| GenBank mRNA | JO579796.1 | - | 3 - 4 | 1395 | 55 | |
More Information
Set Membership: |
|

PLVAP
Genomic Map