Assay Details
| Gene Symbol: PKIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684511.3 | XP_005684568.1 | 2 - 3 | 281 | 76 | |
| XM_018052992.1 | XP_017908481.1 | 5 - 6 | 735 | 76 | ||
| XM_018052993.1 | XP_017908482.1 | 6 - 7 | 846 | 76 | ||
| XM_018052995.1 | XP_017908484.1 | 4 - 5 | 510 | 76 | ||
| XM_018052996.1 | XP_017908485.1 | 4 - 5 | 636 | 76 | ||
| XM_018052997.1 | XP_017908486.1 | 3 - 4 | 510 | 76 | ||
| XM_018052998.1 | XP_017908487.1 | 4 - 5 | 494 | 76 | ||
| XM_018052999.1 | XP_017908488.1 | 5 - 6 | 605 | 76 | ||
| XM_018053000.1 | XP_017908489.1 | 3 - 4 | 424 | 76 | ||
| XM_018053001.1 | XP_017908490.1 | 4 - 5 | 535 | 76 | ||
More Information
Set Membership: |
|

PKIB
Genomic Map