Assay Details
Gene Symbol: LOC102173569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684769.3 | XP_005684826.3 | 1 - 2 | 295 | 95 | |
GenBank mRNA | JP350594.2 | - | 2 - 3 | 275 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
