Assay Details
| Gene Symbol: LOC102173569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684769.3 | XP_005684826.3 | 2 - 3 | 424 | 67 | |
| GenBank mRNA | JP350594.2 | - | 3 - 4 | 404 | 67 | |
More Information
Set Membership: |
|

LOC102173569
Genomic Map