Assay Details
| Gene Symbol: LOC102173569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684769.3 | XP_005684826.3 | 4 - 5 | 907 | 72 | |
| GenBank mRNA | JP350594.2 | - | 5 - 6 | 887 | 72 | |
More Information
Set Membership: |
|

LOC102173569
Genomic Map