Assay Details
| Gene Symbol: LOC102173569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684769.3 | XP_005684826.3 | 5 - 6 | 1279 | 104 | |
| GenBank mRNA | JP350594.2 | - | 6 - 7 | 1259 | 104 | |
More Information
Set Membership: |
|

LOC102173569
Genomic Map