Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687980.3 | XP_005688037.1 | 5 - 6 | 655 | 65 | |
| GenBank mRNA | JO428428.1 | - | 9 - 10 | 2007 | 65 | |
More Information
Set Membership: |
|

STAM
Genomic Map