Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 25 - 26 | 3573 | 59 | |
XM_018059552.1 | XP_017915041.1 | 25 - 26 | 3569 | 59 | ||
XM_018059553.1 | XP_017915042.1 | 25 - 26 | 3573 | 59 | ||
XM_018059554.1 | XP_017915043.1 | 24 - 25 | 3510 | 59 | ||
XM_018059555.1 | XP_017915044.1 | 25 - 26 | 3450 | 59 | ||
XM_018059556.1 | XP_017915045.1 | 23 - 24 | 3384 | 59 | ||
XM_018059557.1 | XP_017915046.1 | 21 - 22 | 3003 | 59 | ||
XM_018059558.1 | XP_017915047.1 | 20 - 21 | 2940 | 59 | ||
XM_018059559.1 | XP_017915048.1 | 20 - 21 | 2895 | 59 | ||
XM_018059560.1 | XP_017915049.1 | 19 - 20 | 2831 | 59 | ||
XM_018059561.1 | XP_017915050.1 | 25 - 26 | 3567 | 59 | ||
XM_018059562.1 | XP_017915051.1 | 25 - 26 | 3567 | 59 | ||
XM_018059564.1 | XP_017915053.1 | 24 - 25 | 3503 | 59 | ||
XM_018059565.1 | XP_017915054.1 | 24 - 25 | 3503 | 59 | ||
GenBank mRNA | KA322253.1 | - | 12 - 13 | 3146 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
