Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 27 - 28 | 3851 | 84 | |
XM_018059552.1 | XP_017915041.1 | 27 - 28 | 3847 | 84 | ||
XM_018059553.1 | XP_017915042.1 | 27 - 28 | 3851 | 84 | ||
XM_018059554.1 | XP_017915043.1 | 26 - 27 | 3788 | 84 | ||
XM_018059555.1 | XP_017915044.1 | 27 - 28 | 3728 | 84 | ||
XM_018059556.1 | XP_017915045.1 | 25 - 26 | 3662 | 84 | ||
XM_018059557.1 | XP_017915046.1 | 23 - 24 | 3281 | 84 | ||
XM_018059558.1 | XP_017915047.1 | 22 - 23 | 3218 | 84 | ||
XM_018059559.1 | XP_017915048.1 | 22 - 23 | 3173 | 84 | ||
XM_018059560.1 | XP_017915049.1 | 21 - 22 | 3109 | 84 | ||
GenBank mRNA | KA322253.1 | - | 10 - 11 | 2868 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
