Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 28 - 29 | 4038 | 76 | |
XM_018059552.1 | XP_017915041.1 | 28 - 29 | 4034 | 76 | ||
XM_018059553.1 | XP_017915042.1 | 28 - 29 | 4038 | 76 | ||
XM_018059554.1 | XP_017915043.1 | 27 - 28 | 3975 | 76 | ||
XM_018059555.1 | XP_017915044.1 | 28 - 29 | 3915 | 76 | ||
XM_018059556.1 | XP_017915045.1 | 26 - 27 | 3849 | 76 | ||
XM_018059557.1 | XP_017915046.1 | 24 - 25 | 3468 | 76 | ||
XM_018059558.1 | XP_017915047.1 | 23 - 24 | 3405 | 76 | ||
XM_018059559.1 | XP_017915048.1 | 23 - 24 | 3360 | 76 | ||
XM_018059560.1 | XP_017915049.1 | 22 - 23 | 3296 | 76 | ||
GenBank mRNA | KA322253.1 | - | 9 - 10 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
