Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 29 - 30 | 4111 | 63 | |
XM_018059552.1 | XP_017915041.1 | 29 - 30 | 4107 | 63 | ||
XM_018059553.1 | XP_017915042.1 | 29 - 30 | 4111 | 63 | ||
XM_018059554.1 | XP_017915043.1 | 28 - 29 | 4048 | 63 | ||
XM_018059555.1 | XP_017915044.1 | 29 - 30 | 3988 | 63 | ||
XM_018059556.1 | XP_017915045.1 | 27 - 28 | 3922 | 63 | ||
XM_018059557.1 | XP_017915046.1 | 25 - 26 | 3541 | 63 | ||
XM_018059558.1 | XP_017915047.1 | 24 - 25 | 3478 | 63 | ||
XM_018059559.1 | XP_017915048.1 | 24 - 25 | 3433 | 63 | ||
XM_018059560.1 | XP_017915049.1 | 23 - 24 | 3369 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
