Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 34 - 35 | 5897 | 86 | |
XM_018059552.1 | XP_017915041.1 | 34 - 35 | 5893 | 86 | ||
XM_018059554.1 | XP_017915043.1 | 33 - 34 | 5834 | 86 | ||
XM_018059555.1 | XP_017915044.1 | 34 - 35 | 5774 | 86 | ||
XM_018059556.1 | XP_017915045.1 | 32 - 33 | 5708 | 86 | ||
XM_018059557.1 | XP_017915046.1 | 30 - 31 | 5327 | 86 | ||
XM_018059558.1 | XP_017915047.1 | 29 - 30 | 5264 | 86 | ||
XM_018059559.1 | XP_017915048.1 | 29 - 30 | 5219 | 86 | ||
XM_018059560.1 | XP_017915049.1 | 28 - 29 | 5155 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
