Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690466.3 | XP_005690523.1 | 4 - 5 | 882 | 66 | |
| XM_005690467.2 | XP_005690524.1 | 4 - 5 | 1307 | 66 | ||
| XM_013970070.2 | XP_013825524.1 | 4 - 5 | 933 | 66 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map