Assay Details
| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062113.1 | XP_017917602.1 | 2 - 3 | 511 | 91 | |
| XM_018062114.1 | XP_017917603.1 | 3 - 4 | 718 | 91 | ||
| XM_018062115.1 | XP_017917604.1 | 2 - 3 | 515 | 91 | ||
| GenBank mRNA | JT745683.1 | - | 3 - 4 | 558 | 91 | |
| KA312779.1 | - | 2 - 3 | 483 | 91 | ||
More Information
Set Membership: |
|

NOL3
Genomic Map