Assay Details
| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062113.1 | XP_017917602.1 | 3 - 4 | 847 | 80 | |
| XM_018062114.1 | XP_017917603.1 | 4 - 5 | 1054 | 80 | ||
| XM_018062115.1 | XP_017917604.1 | 3 - 4 | 839 | 68 | ||
| GenBank mRNA | KA312779.1 | - | 1 - 2 | 147 | 80 | |
More Information
Set Membership: |
|

NOL3
Genomic Map